Tisk…

1-Acetyl-5-indolinesulphonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-acetyl-2,3-dihydro-1H-indole-5-sulphonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetylindoline-5-sulphonyl chloride

OR4396-1G OR4396-250MG OR4396-5G
APOSOR4396-1GEA 1700 CZK
APOSOR4396-1G APOSOR4396-250MG APOSOR4396-5G
1-Acetyl-5-indolinesulphonyl chloride 97%
1-Acetyl-5-indolinesulphonyl chloride
Vzorec: C₁₀H₁₀ClNO₃S
MW: 259.71 g/mol
Bod tání: 172…174 °C
Storage Temperature: Freezer
MDL Number: MFCD07368558
CAS číslo: 52206-05-0
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR