Tisk…

3-Amino-5-cyclopropylpyridine

Dodavatel: Apollo Scientific
Varování

OR42287-1G OR42287-250MG OR42287-5G
APOSOR42287-1GEA 5390 CZK
APOSOR42287-1G APOSOR42287-250MG APOSOR42287-5G
3-Amino-5-cyclopropylpyridine
3-Amino-5-cyclopropylpyridine
Vzorec: C₈H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD19543987
CAS číslo: 1314353-68-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR