Tisk…

Methyl 4-(2-hydroxyethyl)benzoate 95%

Dodavatel: Apollo Scientific
Varování

OR42262-250MG
APOSOR42262-250MGEA 3110 CZK
APOSOR42262-250MG
Methyl 4-(2-hydroxyethyl)benzoate 95%
Methyl 4-(2-hydroxyethyl)benzoate
Vzorec: C₁₀H₁₂O₃
MW: 180.2 g/mol
Bod varu: 137 °C (2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD24447574
CAS číslo: 46190-45-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR