Tisk…

3-Amino-8-methoxyquinoline dihydrochloride

Dodavatel: Apollo Scientific

OR42241-1G
APOSOR42241-1GEA 17990 CZK
APOSOR42241-1G
3-Amino-8-methoxyquinoline dihydrochloride
3-Amino-8-methoxyquinoline dihydrochloride
Vzorec: C₁₀H₁₀N₂O·2ClH
Storage Temperature: Ambient
MDL Number: MFCD18375326
CAS číslo: 1266226-29-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR