Tisk…

2-Methyl-4-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR42154-1G OR42154-25G OR42154-5G
APOSOR42154-1GEA 744 CZK
APOSOR42154-1G APOSOR42154-25G APOSOR42154-5G
2-Methyl-4-pyridineboronic acid pinacol ester
2-Methyl-4-pyridineboronic acid pinacol ester
Vzorec: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Bod tání: 66…71 °C
Storage Temperature: Freezer
MDL Number: MFCD08061590
CAS číslo: 660867-80-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR