Tisk…

2-Bromo-4-cyclopropylaniline 96%

Dodavatel: Apollo Scientific
Varování

OR40777-1G OR40777-250MG OR40777-5G
APOSOR40777-1GEA 14060 CZK
APOSOR40777-1G APOSOR40777-250MG APOSOR40777-5G
2-Bromo-4-cyclopropylaniline 96%
2-Bromo-4-cyclopropylaniline
Vzorec: C₉H₁₀BrN
MW: 212.086 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21099997
CAS číslo: 1353857-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR