Tisk…

(R)-1-Boc-3-Ethylpiperazine 95%

Dodavatel: Apollo Scientific
Varování

OR40701-250MG OR40701-1G
APOSOR40701-250MGEA 787 CZK
APOSOR40701-250MG APOSOR40701-1G
(R)-1-Boc-3-Ethylpiperazine 95%
(R)-1-Boc-3-Ethylpiperazine
Vzorec: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07772111
CAS číslo: 438050-08-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR