Tisk…

Ethyl-2-chloropyrimidine-5-carboxylate

Dodavatel: Apollo Scientific
Varování

OR40379-5G OR40379-1G OR40379-25G
APOSOR40379-5GEA 1780 CZK
APOSOR40379-5G APOSOR40379-1G APOSOR40379-25G
Ethyl-2-chloropyrimidine-5-carboxylate
Ethyl-2-chloropyrimidine-5-carboxylate
Vzorec: C₇H₇ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09863164
CAS číslo: 89793-12-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR