Tisk…

2-Hydroxy-4-pyridineboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

OR40294-1G OR40294-250MG OR40294-5G
APOSOR40294-1GEA 7770 CZK
APOSOR40294-1G APOSOR40294-250MG APOSOR40294-5G
2-Hydroxy-4-pyridineboronic acid pinacol ester
2-Hydroxy-4-pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₆BNO₃
MW: 221.06
Storage Temperature: Ambient
MDL Number: MFCD11223287
CAS číslo: 1256358-90-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR