Tisk…

3,5-Dimethylbenzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR40202-100G OR40202-250G OR40202-500G
APOSOR40202-100GEA 2900 CZK
APOSOR40202-100G APOSOR40202-250G APOSOR40202-500G
3,5-Dimethylbenzoyl chloride
3,5-Dimethylbenzoyl chloride
Vzorec: C₉H₉ClO
MW: 168.62
Bod varu: 127 °C/20mmHg
Hustota: 1.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02313742
CAS číslo: 6613-44-1
EINECS: 413-010-9
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR