Tisk…

2-Amino-5-methoxypyrazine 98%

Dodavatel: Apollo Scientific

OR40076-1G
APOSOR40076-1GEA 2650 CZK
APOSOR40076-1G
2-Amino-5-methoxypyrazine 98%
2-Amino-5-methoxypyrazine
Vzorec: C₅H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD09909647
CAS číslo: 54013-07-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR