Tisk…

4-Bromo-2-nitrobenzonitrile 98%

Dodavatel: Apollo Scientific
Varování

OR400603-1G OR400603-5G OR400603-10G
APOSOR400603-1GEA 1620 CZK
APOSOR400603-1G APOSOR400603-5G APOSOR400603-10G
4-Bromo-2-nitrobenzonitrile 98%
4-Bromo-2-nitrobenzonitrile
Vzorec: C₇H₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00087225
CAS číslo: 79603-03-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR