Tisk…

2-Phenylethylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Phenethylboronic acid

OR4006-1G OR4006-5G
APOSOR4006-1GEA 622 CZK
APOSOR4006-1G APOSOR4006-5G
2-Phenylethylboronic acid
2-Phenylethylboronic acid
Vzorec: C₈H₁₁BO₂
MW: 149.98 g/mol
Bod tání: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01631226
CAS číslo: 34420-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR