Tisk…

2-Hydroxypyrazine 95%

Dodavatel: Apollo Scientific
Varování

OR40046-1G OR40046-5G
APOSOR40046-1GEA 622 CZK
APOSOR40046-1G APOSOR40046-5G
2-Hydroxypyrazine 95%
2-Hydroxypyrazine
Vzorec: C₄H₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00233966
CAS číslo: 6270-63-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR