Tisk…

2-Bromo-4-(tert-octyl)phenol

Dodavatel: Apollo Scientific
Varování

OR40033-25G OR40033-5G OR40033-100G
APOSOR40033-25GEA 870 CZK
APOSOR40033-25G APOSOR40033-5G APOSOR40033-100G
2-Bromo-4-(tert-octyl)phenol
2-Bromo-4-(tert-octyl)phenol
Vzorec: C₁₄H₂₁BrO
Storage Temperature: Ambient
MDL Number: MFCD09907053
CAS číslo: 57835-35-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR