Tisk…

4-Bromo-5-methyl-2-nitroanisole 95+%

Dodavatel: Apollo Scientific

Synonyms: 1-Bromo-4-methoxy-2-methyl-5-nitrobenzene

OR400208-1G OR400208-5G
APOSOR400208-1GEA 1740 CZK
APOSOR400208-1G APOSOR400208-5G
4-Bromo-5-methyl-2-nitroanisole 95+%
4-Bromo-5-methyl-2-nitroanisole
Vzorec: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23703052
CAS číslo: 89978-56-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR