Tisk…

4-(Bromomethyl)phenyl acetate 97%, tech

Dodavatel: Apollo Scientific
Nebezpečí

OR400140-1G OR400140-5G
APOSOR400140-1GEA 6220 CZK
APOSOR400140-1G APOSOR400140-5G
4-(Bromomethyl)phenyl acetate 97%, tech
4-(Bromomethyl)phenyl acetate
Vzorec: C₉H₉BrO₂
Storage Temperature: Freezer
MDL Number: MFCD17013576
CAS číslo: 52727-95-4
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR