Tisk…

7-Bromobenzofuran-3(2H)-one

Dodavatel: Apollo Scientific

OR400051-1G OR400051-250MG
APOSOR400051-1GEA 7870 CZK
APOSOR400051-1G APOSOR400051-250MG
7-Bromobenzofuran-3(2H)-one
7-Bromobenzofuran-3(2H)-one
Vzorec: C₈H₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD03160589
CAS číslo: 519018-52-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR