Tisk…

4-(trans-2-Carboxyvinyl)phenylboronic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: (E)-3-(4-Boronophenyl)acrylic acid

OR3953-1G OR3953-5G OR3953-25G
APOSOR3953-1GEA 2320 CZK
APOSOR3953-1G APOSOR3953-5G APOSOR3953-25G
4-(trans-2-Carboxyvinyl)phenylboronic acid 96%
4-(trans-2-Carboxyvinyl)phenylboronic acid
Vzorec: C₉H₉BO₄
MW: 191.9764 g/mol
Bod tání: 222…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075704
CAS číslo: 159896-15-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR