Tisk…

Methyl 4-methoxy-2-naphthoate ≥98%

Dodavatel: Apollo Scientific

OR370022-1G OR370022-250MG
APOSOR370022-1GEA 39020 CZK
APOSOR370022-1G APOSOR370022-250MG
Methyl 4-methoxy-2-naphthoate ≥98%
Methyl 4-methoxy-2-naphthoate
Vzorec: C₁₃H₁₂O₃
MW: 216.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181453
CAS číslo: 33295-47-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR