Tisk…

5-Chloro-2-methylindole

Dodavatel: Apollo Scientific

OR3699-5G
APOSOR3699-5GEA 1620 CZK
APOSOR3699-5G
5-Chloro-2-methylindole
5-Chloro-2-methylindole
Vzorec: C₉H₈ClN
MW: 165.62
Bod tání: 129
Storage Temperature: Ambient
MDL Number: MFCD00005619
CAS číslo: 1075-35-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR