Tisk…

Fmoc-p-azido-D-Phe-OH 99%

Dodavatel: Apollo Scientific

OR36432-1G OR36432-250MG
APOSOR36432-1GEA 15710 CZK
APOSOR36432-1G APOSOR36432-250MG
Fmoc-p-azido-D-Phe-OH 99%
Fmoc-p-azido-D-Phe-OH
Vzorec: C₂₄H₂₀N₄O₄
MW: 428.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11041252
CAS číslo: 1391586-30-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR