Tisk…

1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%

Dodavatel: Apollo Scientific
Varování

OR360855-250MG OR360855-1G
APOSOR360855-250MGEA 2440 CZK
APOSOR360855-250MG APOSOR360855-1G
1-tert-Butylpyrazole-4-boronic acid, pinacol ester 97%
1-tert-Butylpyrazole-4-boronic acid, pinacol ester
Vzorec: C₁₃H₂₃BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16660302
CAS číslo: 1256359-15-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR