Tisk…

(3-([(tert-Butoxycarbonyl)amino]methyl)pyridin-4-yl)boronic acid 95%

Dodavatel: Apollo Scientific
Varování

OR360584-100MG OR360584-1G OR360584-250MG
APOSOR360584-100MGEA 7250 CZK
APOSOR360584-100MG APOSOR360584-1G APOSOR360584-250MG
(3-([(tert-Butoxycarbonyl)amino]methyl)pyridin-4-yl)boronic acid 95%
(3-([(tert-Butoxycarbonyl)amino]methyl)pyridin-4-yl)boronic acid
Vzorec: C₁₁H₁₇BN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD05664023
CAS číslo: 433969-29-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR