Tisk…

3-Acetamidophenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3'-Acetamido-2-bromoacetophenone

OR350581-1G OR350581-250MG OR350581-5G
APOSOR350581-1GEA 8070 CZK
APOSOR350581-1G APOSOR350581-250MG APOSOR350581-5G
3-Acetamidophenacyl bromide
3-Acetamidophenacyl bromide
Vzorec: C₁₀H₁₀BrNO₂
MW: 256.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07364400
CAS číslo: 30095-56-8
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR