Tisk…

7-Bromo-5-methylbenzo[e][1,2,4]triazin-3-amine ≥98%

Dodavatel: Apollo Scientific

OR350428-1G
APOSOR350428-1GEA 17030 CZK
APOSOR350428-1G
7-Bromo-5-methylbenzo[e][1,2,4]triazin-3-amine ≥98%
7-Bromo-5-methylbenzo[e][1,2,4]triazin-3-amine
Vzorec: C₈H₇BrN₄
MW: 239.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031506
CAS číslo: 867330-26-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR