Tisk…

1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester

Dodavatel: Apollo Scientific

OR350388-1G
APOSOR350388-1GEA 3060 CZK
APOSOR350388-1G
1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester
1-Tosyl-1,2,3,6-tetrahydropyridine-4-boronic acid, pinacol ester
Vzorec: C₁₈H₂₆BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD17926518
CAS číslo: 1256360-46-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR