Tisk…

Sodium-6,7-dihydroxynaphthalene-2-sulphonate

Dodavatel: Apollo Scientific

Synonyms: 2,3-Dihydroxynaphthalene-6-sulphonic acid Monosodium Salt, DS acid Monosodium Salt, Sodium 2,3-dihydroxynaphthalene-6-sulphonate, Monosodium 6,7-Dihydroxynaphthalene-2-sulphonate

OR350345-100G
APOSOR350345-100GEA 1780 CZK
APOSOR350345-100G
Sodium-6,7-dihydroxynaphthalene-2-sulphonate
Sodium-6,7-dihydroxynaphthalene-2-sulphonate
Vzorec: C₁₀H₈O₅S
MW: 262.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004074
CAS číslo: 135-53-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR