Tisk…

Pyridoxal-5'-phosphate monohydrate 98%

Dodavatel: Apollo Scientific

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

OR350314-100G OR350314-25G OR350314-5G
APOSOR350314-100GEA 8700 CZK
APOSOR350314-100G APOSOR350314-25G APOSOR350314-5G
Pyridoxal-5'-phosphate monohydrate 98%
Pyridoxal-5'-phosphate monohydrate
Vzorec: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS číslo: 41468-25-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR