Tisk…

[4-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%

Dodavatel: Apollo Scientific
Varování

OR350198-1G OR350198-5G
APOSOR350198-1GEA 1950 CZK
APOSOR350198-1G APOSOR350198-5G
[4-(1H-Pyrazol-1-yl)phenyl]boronic acid 98%
[4-(1H-Pyrazol-1-yl)phenyl]boronic acid
Vzorec: C₉H₉BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06739055
CAS číslo: 891270-35-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR