Tisk…

5-Benzyloxy-2-methylpyridine 95+%

Dodavatel: Apollo Scientific

Synonyms: 5-(Benzyloxy)-2-methylpyridine

OR346578-1G
APOSOR346578-1GEA 17760 CZK
APOSOR346578-1G
5-Benzyloxy-2-methylpyridine 95+%
5-Benzyloxy-2-methylpyridine
Vzorec: C₁₃H₁₃NO
MW: 199.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15523327
CAS číslo: 63793-98-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR