Tisk…

6-Chloro-5-methyl-pyridine-3-sulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR346310-1G
APOSOR346310-1GEA 0 CZK
APOSOR346310-1G
6-Chloro-5-methyl-pyridine-3-sulphonyl chloride
6-Chloro-5-methyl-pyridine-3-sulphonyl chloride
Vzorec: C₆H₅Cl₂NO₂S
Bod tání: 56…57 °C
Storage Temperature: Ambient
MDL Number: MFCD09881190
CAS číslo: 37105-10-5
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR