Tisk…

(1,4-Diphenyl-1H-pyrrol-3-yl)-methanol

Dodavatel: Apollo Scientific

OR346308-1G
APOSOR346308-1GEA 0 CZK
APOSOR346308-1G
(1,4-Diphenyl-1H-pyrrol-3-yl)-methanol
(1,4-Diphenyl-1H-pyrrol-3-yl)-methanol
Vzorec: C₁₇H₁₅NO
MW: 249.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21362495
CAS číslo: 1355334-89-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR