Tisk…

Benzofuran-3-carboxylic acid methyl ester

Dodavatel: Apollo Scientific
Varování

OR346248-1G OR346248-500MG OR346248-5G
APOSOR346248-1GEA 0 CZK
APOSOR346248-1G APOSOR346248-500MG APOSOR346248-5G
Benzofuran-3-carboxylic acid methyl ester
Benzofuran-3-carboxylic acid methyl ester
Vzorec: C₁₀H₈O₃
MW: 176.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144245
CAS číslo: 4687-24-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR