Tisk…

3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester

Dodavatel: Apollo Scientific

OR346203-1G OR346203-500MG
APOSOR346203-1GEA 0 CZK
APOSOR346203-1G APOSOR346203-500MG
3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester
3-Cyclopropyl-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester
Vzorec: C₈H₁₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09701787
CAS číslo: 1018125-29-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR