Tisk…

C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine

Dodavatel: Apollo Scientific

OR346073-1G OR346073-5G
APOSOR346073-1GEA 0 CZK
APOSOR346073-1G APOSOR346073-5G
C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine
C-(3-Phenyl-[1,2,4]oxadiazol-5-yl)methylamine
Vzorec: C₉H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD06166274
CAS číslo: 90564-77-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR