Tisk…

5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one

Dodavatel: Apollo Scientific

OR345716-1G
APOSOR345716-1GEA 5290 CZK
APOSOR345716-1G
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one
Vzorec: C₅H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18427081
CAS číslo: 4722-76-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR