Tisk…

4-Chloro-2-methyl-7-azaindole 95%

Dodavatel: Apollo Scientific

OR345316-1G OR345316-250MG OR345316-10G
APOSOR345316-1GEA 2820 CZK
APOSOR345316-1G APOSOR345316-250MG APOSOR345316-10G
4-Chloro-2-methyl-7-azaindole 95%
4-Chloro-2-methyl-7-azaindole
Vzorec: C₈H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD12924600
CAS číslo: 307951-53-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR