Tisk…

5-Acetyl-2-methoxypyridine 98%

Dodavatel: Apollo Scientific

OR345295-25G
APOSOR345295-25GEA 6750 CZK
APOSOR345295-25G
5-Acetyl-2-methoxypyridine 98%
5-Acetyl-2-methoxypyridine
Vzorec: C₈H₉NO₂
MW: 151.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664350
CAS číslo: 213193-32-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR