Tisk…

2-Phenethylpyrazine 95%

Dodavatel: Apollo Scientific

OR33584-1G
APOSOR33584-1GEA 7670 CZK
APOSOR33584-1G
2-Phenethylpyrazine 95%
2-Phenethylpyrazine
Vzorec: C₁₂H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD16140369
CAS číslo: 91391-83-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR