Tisk…

Ethyl-5-methylpicolinate 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 5-methylpicolinate, Ethyl 5-methylpyridine-2-carboxylate

OR33564-1G OR33564-5G OR33564-25G
APOSOR33564-1GEA 5130 CZK
APOSOR33564-1G APOSOR33564-5G APOSOR33564-25G
Ethyl-5-methylpicolinate 97%
Ethyl-5-methylpicolinate
Vzorec: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD10566273
CAS číslo: 55876-82-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR