Tisk…

2-(4-Chlorophenyl)-2-oxoethyl acetate 95%

Dodavatel: Apollo Scientific

OR33513-1G OR33513-5G
APOSOR33513-1GEA 5130 CZK
APOSOR33513-1G APOSOR33513-5G
2-(4-Chlorophenyl)-2-oxoethyl acetate 95%
2-(4-Chlorophenyl)-2-oxoethyl acetate
Vzorec: C₁₀H₉ClO₃
MW: 212.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00662320
CAS číslo: 39561-82-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR