Tisk…

(2E)-2-[(Thiophen-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Dodavatel: Apollo Scientific

OR33365-1G
APOSOR33365-1GEA 44640 CZK
APOSOR33365-1G
(2E)-2-[(Thiophen-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2E)-2-[(Thiophen-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Vzorec: C₁₂H₁₃NOS
Storage Temperature: Ambient
MDL Number: MFCD06797029
CAS číslo: 82955-19-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR