Tisk…

5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%

Dodavatel: Apollo Scientific

OR33215-1G OR33215-5G
APOSOR33215-1GEA 9560 CZK
APOSOR33215-1G APOSOR33215-5G
5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
5-Chloro-1-(4-chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Vzorec: C₁₃H₉Cl₂NO₃
MW: 298.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00140291
CAS číslo: 339024-25-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR