Tisk…

2-Cyclopropyl-5-methoxy-1-benzofuran-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR33053-1G
APOSOR33053-1GEA 24900 CZK
APOSOR33053-1G
2-Cyclopropyl-5-methoxy-1-benzofuran-3-carbaldehyde 95%
2-Cyclopropyl-5-methoxy-1-benzofuran-3-carbaldehyde
Vzorec: C₁₃H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11553025
CAS číslo: 1092352-26-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR