Tisk…

1-(Morpholin-4-yl)-3-phenoxypropan-2-ol 95%

Dodavatel: Apollo Scientific

OR33045-1G
APOSOR33045-1GEA 24900 CZK
APOSOR33045-1G
1-(Morpholin-4-yl)-3-phenoxypropan-2-ol 95%
1-(Morpholin-4-yl)-3-phenoxypropan-2-ol
Vzorec: C₁₃H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00450856
CAS číslo: 5296-26-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR