Tisk…

3-[3-(Dimethylamino)prop-2-enoyl]benzonitrile, tech

Dodavatel: Apollo Scientific

OR32939-1G
APOSOR32939-1GEA 44640 CZK
APOSOR32939-1G
3-[3-(Dimethylamino)prop-2-enoyl]benzonitrile, tech
3-[3-(Dimethylamino)prop-2-enoyl]benzonitrile
Vzorec: C₁₂H₁₂N₂O
MW: 200,236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00111583
CAS číslo: 179055-13-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR