Tisk…

3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%

Dodavatel: Apollo Scientific

OR32781-1G OR32781-5G
APOSOR32781-1GEA 4630 CZK
APOSOR32781-1G APOSOR32781-5G
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
3-Hydroxy-2-phenyl-4(1H)-quinolinone
Vzorec: C₁₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00451324
CAS číslo: 31588-18-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR