Tisk…

7-Bromo-1H-indole-3-carboxaldehyde

Dodavatel: Apollo Scientific
Varování

OR3269-1G
APOSOR3269-1GEA 1660 CZK
APOSOR3269-1G
7-Bromo-1H-indole-3-carboxaldehyde
7-Bromo-1H-indole-3-carboxaldehyde
Vzorec: C₉H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD06657151
CAS číslo: 115666-21-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR