Tisk…

(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one, tech

Dodavatel: Apollo Scientific

OR32679-1G
APOSOR32679-1GEA 44640 CZK
APOSOR32679-1G
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one, tech
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one
Vzorec: C₁₀H₁₂BrNOS
MW: 274,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03618051
CAS číslo: 865657-77-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR